missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Naphthyl)propionic acid, 96%

Product Code. 15466657
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466657 1 g 1g
15476657 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466657 Supplier Thermo Scientific Alfa Aesar Supplier No. H33795.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3243-42-3 | C13H12O2 | 200.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3243-42-3
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00033058
InChI Key PRLKVVMRQFFIOQ-UHFFFAOYSA-N
Synonym 3-1-naphthyl propionic acid, 3-naphthalen-1-yl propanoic acid, 1-naphthalenepropanoic acid, 1-naphthalenepropionic acid, 3-1-naphthyl-propionic acid, 3-1-naphthyl propanoic acid, 3-naphth-1-yl propanoic acid, 3-naphthalen-1-yl-propionic acid, 1-naphthalenepropanoicacid, 3-naphthylpropanoic acid
PubChem CID 95254
IUPAC Name 3-naphthalen-1-ylpropanoic acid
SMILES OC(=O)CCC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 155°C to 159°C
Quantity 1 g
Formula Weight 200.24
Percent Purity 96%
Chemical Name or Material 3-(1-Naphthyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.