missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-1,2,4-Triazol-1-yl)phenyl]methanol, ≥95%, Thermo Scientific™

Product Code. 11370607
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11370607 250 mg Amber Glass Bottle 250mg
10653445 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11370607 Supplier Thermo Scientific Maybridge Supplier No. CC38409CB
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 868755-55-9
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.19
MDL Number MFCD08271897
InChI Key KUSZGGXHHOYFHS-UHFFFAOYSA-N
Synonym 3-1h-1,2,4-triazol-1-yl phenyl methanol, 3-1,2,4-triazol-1-yl phenyl methanol, 3-1,2,4-triazolyl phenyl methan-1-ol
PubChem CID 20485344
IUPAC Name [3-(1H-1,2,4-triazol-1-yl)phenyl]methanol
SMILES OCC1=CC(=CC=C1)N1C=NC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material [3-(1H-1, 2, 4-triazol-1-yl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.