missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10081384
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10081384 250 mg 250mg
10386324 1 g 1g
10265314 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081384 Supplier Thermo Scientific Maybridge Supplier No. CC43204CB
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 102432-05-3
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD08271917
InChI Key JMKQKKQFLYPQDD-UHFFFAOYSA-N
PubChem CID 10397537
IUPAC Name 3-(imidazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC(=C1)CN2C=CN=C2)C=O

Tekniske data

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 3-1h-imidazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-imidazol-1-ylmethyl, 3-imidazol-1-ylmethyl benzaldehyde, 3-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde, 3-1h-imidazol-1-ylmethyl, acmc-1c5ro, 3-imidazolylmethyl benzaldehyde
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 3-(1H-imidazol-1-ylmethyl)benzaldehyde
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.