missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 10356234
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10356234 1 g 1g
10275984 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10356234 Supplier Thermo Scientific Maybridge Supplier No. CC56113DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 562803-76-3
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD08572164
InChI Key XKFHMAZBNKKNMT-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-ylmethyl benzylamine, 3-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-3-aminomethyl benzyl-1h-pyrazole, 3-pyrazol-1-ylmethyl phenyl methanamine, 1-3-pyrazol-1-ylmethyl phenyl methanamine, 3-pyrazolylmethyl phenyl methylamine, 3-1h-pyrazol-1-yl methyl benzylamine, 3-1h-pyrazole-1-ylmethyl benzenemethanamine, benzenemethanamine,3-1h-pyrazol-1-ylmethyl, 3-1h-pyrazol-1-yl methyl phenyl methanamine
PubChem CID 16640564
IUPAC Name [3-(pyrazol-1-ylmethyl)phenyl]methanamine
SMILES C1=CC(=CC(=C1)CN)CN2C=CC=N2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 1 g
Formula Weight 187.24
Percent Purity 90%
Chemical Name or Material 3-(1H-Pyrazol-1-ylmethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.