missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10576595
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10576595 5 g 5g
10212832 250 mg 250mg
10603213 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10576595 Supplier Thermo Scientific Maybridge Supplier No. CC21909DE
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 83140-94-7
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD03086142
InChI Key QQXDXYAGEXWXQU-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl phenyl methanol, 3-pyrrol-1-yl phenyl methanol, 3-1-pyrrolyl benzyl alcohol, 3-pyrrol-1-ylphenyl methanol, 3-pyrrolylphenyl methan-1-ol, 3-pyrrol-1-yl phenylmethanol, 3-1h-pyrrol-1-yl phenylmethanol, benzenemethanol, 3-1h-pyrrol-1-yl
PubChem CID 2776531
IUPAC Name (3-pyrrol-1-ylphenyl)methanol
SMILES C1=CN(C=C1)C2=CC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [3-(1H-Pyrrol-1-yl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.