missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-5-yl)phenol, 97%

Product Code. p-7030003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483707 250 mg 250mg
11481800 1 g 1g
11491800 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15483707 Supplier Thermo Scientific Alfa Aesar Supplier No. H27529.MD
Add to basket
This item is not returnable. View return policy

CAS: 96859-34-6 | C7H6N4O | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96859-34-6
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD06797247
InChI Key IZORRBUQWFSCII-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-5-yl phenol, 5-3-hydroxyphenyl tetrazole, 3-2h-tetrazol-5-yl phenol, 3-2h-1,2,3,4-tetrazol-5-yl phenol, 3-1h-1,2,3,4-tetrazol-5-yl phenol, m-tetrazolylphenol, 3-2h-tetrazol-5-yl-phenol, 5-3-hydroxyphenyl-1h-tetrazole
PubChem CID 13455763
SMILES OC1=CC=CC(=C1)C1=NNN=N1

Specifications

Melting Point ∽220°C (decomposition)
Quantity 250 mg
IUPAC Name 3-(2H-tetrazol-5-yl)phenol
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 3-(1H-Tetrazol-5-yl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.