missing translation for 'onlineSavingsMsg'
Learn More

3-(2,3-Dimethoxyphenyl)propionic acid, 96%, Thermo Scientific™

Product Code. 15427447
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427447 10 g 10g
15417447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427447 Supplier Thermo Scientific Alfa Aesar Supplier No. H34491.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10538-48-4
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00029821
InChI Key QPHAUUNQQLRYCJ-UHFFFAOYSA-N
PubChem CID 314929
IUPAC Name 3-(2,3-dimethoxyphenyl)propanoic acid
SMILES COC1=CC=CC(=C1OC)CCC(=O)O

Specifications

Melting Point 58°C to 60°C
Quantity 10 g
Synonym 3-2,3-dimethoxyphenyl propanoic acid, benzenepropanoic acid, 2,3-dimethoxy, 3-2,3-dimethoxyphenyl propionic acid, 2,3-dimethoxy-benzenepropanoic acid, benzenepropanoic acid,2,3-dimethoxy, acmc-1bqt5, 2.3-dimethoxyhydrozimtsaure, 3-2,3-dimethoxyphenyl propionicacid, 3-2,3-dimethoxyphenyl-propanoic acid, 3-2,3-dimethoxy-phenyl-propionic acid
Formula Weight 210.23
Percent Purity 96%
Chemical Name or Material 3-(2,3-Dimethoxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.