missing translation for 'onlineSavingsMsg'
Learn More

3-(2,4-Dichlorophenyl)propanoic acid, 97%, Thermo Scientific™

Product Code. 10712937
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10712937 10 g 10g
10243142 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712937 Supplier Thermo Scientific Maybridge Supplier No. SEW04678EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 55144-92-8
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.061
MDL Number MFCD00275605
InChI Key HLNPVFSCAMKIOD-UHFFFAOYSA-N
PubChem CID 2740915
IUPAC Name 3-(2,4-dichlorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)CCC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula Cl2C6H3CH2CH2CO2H
Quantity 10 g
Synonym 3-2,4-dichlorophenyl propanoic acid, 3-2,4-dichlorophenyl propionic acid, 2,4-dichlorohydrocinnamic acid, 2,4-dichloro-benzenepropanoic acid, benzenepropanoic acid, 2,4-dichloro, 3-2,4-dichloro-phenyl-propionic acid, maybridge4_004135, acmc-1b21h, ksc603c0n, benzenepropanoic acid,2,4-dichloro
Formula Weight 219.07
Percent Purity 97%
Chemical Name or Material 3-(2, 4-Dichlorophenyl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.