missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2,5-Dimethoxyphenyl)-1H-pyrazole

Product Code. 11410160
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410160 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11410160 Supplier Alfa Aesar™ Supplier No. H50636.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 181122-45-2
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.229
MDL Number MFCD02091528
InChI Key JEFGLUGSZPRTEJ-UHFFFAOYSA-N
Synonym 3-2,5-dimethoxyphenyl-1h-pyrazole, 5-2,5-dimethoxyphenyl-1h-pyrazole, 3-2,5-dimethoxyphenyl pyrazole, 1,4-dimethoxy-2-pyrazol-3-ylbenzene, acmc-1byef
PubChem CID 18801454
IUPAC Name 5-(2,5-dimethoxyphenyl)-1H-pyrazole
SMILES COC1=CC(=C(C=C1)OC)C2=CC=NN2

Specifications

Quantity 1 g
Formula Weight 204.23
Chemical Name or Material 3-(2,5-Dimethoxyphenyl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.