missing translation for 'onlineSavingsMsg'
Learn More

3-(2,5-Dimethoxyphenyl)propionic acid, 96%

Product Code. 15436397
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436397 25 g 25g
15416397 1 g 1g
15426397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15436397 Supplier Thermo Scientific Alfa Aesar Supplier No. H33566.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10538-49-5 | C11H14O4 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10538-49-5
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00060325
InChI Key JENQUCZZZGYHRW-UHFFFAOYSA-N
PubChem CID 66343
IUPAC Name 3-(2,5-dimethoxyphenyl)propanoic acid
SMILES COC1=CC=C(OC)C(CCC(O)=O)=C1

Specifications

Melting Point 61°C to 66°C
Quantity 25 g
Synonym 3-2,5-dimethoxyphenyl propanoic acid, 3-2,5-dimethoxyphenyl propionic acid, benzenepropanoic acid, 2,5-dimethoxy, 2,5-dimethoxyhydrocinnamic acid, 2,5-dimethoxybenzenepropanoic acid, benzenepropanoic acid,2,5-dimethoxy, maybridge1_000808, acmc-1bo6n, ksc503q0f, 2,5-dimethoxy-benzenepropanoic acid
Formula Weight 210.23
Percent Purity 96%
Chemical Name or Material 3-(2,5-Dimethoxyphenyl)propionic acid

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.