missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Bromophenyl)-1H-pyrazole, 97%

Product Code. 15447347
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447347 10 g 10g
15437347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447347 Supplier Thermo Scientific Alfa Aesar Supplier No. H34406.09
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 114382-20-6 | C9H7BrN2 | 223.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114382-20-6
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD01940435
InChI Key VDHTYVJAQNZWTD-UHFFFAOYSA-N
Synonym 3-2-bromophenyl-1h-pyrazole, 3-2-bromophenyl pyrazole, 5-2-bromophenyl-1h-pyrazole, 1h-pyrazole,3-2-bromophenyl, 3-2-bromo-phenyl-1h-pyrazole, 3-2-bromophenyl-2h-pyrazole, 3-o-bromophenylpyrazole, acmc-1c4z7
PubChem CID 2735612
IUPAC Name 5-(2-bromophenyl)-1H-pyrazole
SMILES C1=CC=C(C(=C1)C2=CC=NN2)Br

Specifications

Melting Point 130°C to 132°C
Quantity 10 g
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 3-(2-Bromophenyl)-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.