missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Bromophenyl)-5-methyl-1,2,4-oxadiazole, ≥97%, Thermo Scientific™

Product Code. 10075912
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10075912 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10075912 Supplier Thermo Scientific Maybridge Supplier No. CC47910DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 859851-04-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H7BrN2O
MDL Number MFCD07772874
Quantity 1 g
Synonym 3-2-bromophenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-3-bromophenyl-5-methyl, 3-2-bromophenyl-5-methyl-1,2,4 oxadiazole
InChI Key WLLDZYVHNSXWSY-UHFFFAOYSA-N
SMILES CC1=NC(=NO1)C1=CC=CC=C1Br
IUPAC Name 3-(2-bromophenyl)-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol) 239.07
PubChem CID 7164661
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 3-(2-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.