missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-3,4-dimethoxyphenyl)acrylic acid, 97%, Thermo Scientific™

Product Code. 10314802
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10314802 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10314802 Supplier Thermo Scientific Maybridge Supplier No. BTB09238EA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 99854-17-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H10ClO4
MDL Number MFCD00084947
Quantity 10 g
Synonym 3-2-chloro-3,4-dimethoxyphenyl acrylic acid, 2-chloro-3,4-dimethoxycinnamic acid, 2e-3-2-chloro-3,4-dimethoxyphenyl prop-2-enoic acid, e-3-2-chloro-3,4-dimethoxyphenyl acrylic acid, e-3-2-chloro-3,4-dimethoxy-phenyl-acrylic acid, e-3-2-chloro-3,4-dimethoxyphenyl prop-2-enoic acid, 2e-3-2-chloro-3,4-dimethoxyphenyl-2-propenoic acid
InChI Key JIPTVEJKXDVECY-GQCTYLIASA-M
SMILES COC1=CC=C(\C=C\C([O-])=O)C(Cl)=C1OC
IUPAC Name (E)-3-(2-chloro-3,4-dimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 241.65
PubChem CID 5708524
Formula Weight 242.66
Percent Purity 97%
Chemical Name or Material 3-(2-Chloro-3, 4-dimethoxyphenyl)acrylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.