missing translation for 'onlineSavingsMsg'
Learn More

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide, 97%, Thermo Scientific™

Product Code. 10533232
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10533232 1 g Amber Glass Bottle 1g
10591112 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533232 Supplier Thermo Scientific Maybridge Supplier No. RF00732DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Kemiska identifierare

CAS 4415-11-6
Molecular Formula C11H8ClFN2O2
Molecular Weight (g/mol) 254.65
MDL Number MFCD00173954
InChI Key PPKCMGUTHICRLU-UHFFFAOYSA-N
Synonym 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxamide, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxamide, maybridge1_006010, 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4carboxamide, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxamide, 3-6-chloro-2-fluorophenyl-5-methylisoxazole-4-carboxamide, 4-isoxazolecarboxamide,3-2-chloro-6-fluorophenyl-5-methyl
PubChem CID 2736583

Specifikationer

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 254.65
Percent Purity 97%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.