missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid, TRC

Product Code. 30353655
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353655 10 g -
30368122 1 g 1g
30362079 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353655 Supplier Toronto Research Chemicals Supplier No. TRCC36647510G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3919-74-2
Molecular Formula C11 H7 Cl F N O3
Molecular Weight (g/mol) 255.63
Synonym 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid, Flucloxacillin Sodium Imp. D (EP)
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES Cc1onc(c1C(=O)O)c2c(F)cccc2Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H7ClFNO3/c1-5-8(11(15)16)10(14-17-5)9-6(12)3-2-4-7(9)13/h2-4H,1H3,(H,15,16)
Formula Weight 255.0098
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.