CAS RN 3919-74-2
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 99%
CAS: 3919-74-2 Molecular Formula: C11H7ClFNO3 Molecular Weight (g/mol): 255.63 MDL Number: MFCD00055659 InChI Key: REIPZLFZMOQHJT-UHFFFAOYSA-N Synonym: 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid,3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid,fcimic acid,4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl,3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid,3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid,3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid,pubchem8772,acmc-1ckjh,3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid PubChem CID: 77520 IUPAC Name: 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid SMILES: CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl
3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™
CAS: 3919-74-2 Molecular Formula: C11H7ClFNO3 Molecular Weight (g/mol): 255.63 MDL Number: MFCD00055659 InChI Key: REIPZLFZMOQHJT-UHFFFAOYSA-N Synonym: 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid,3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid,fcimic acid,4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl,3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid,3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid,3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid,pubchem8772,acmc-1ckjh,3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid PubChem CID: 77520 IUPAC Name: 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid SMILES: CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid, TRC
CAS: 3919-74-2 Molecular Formula: C11 H7 Cl F N O3 Molecular Weight (g/mol): 255.63 Synonym: 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid,Flucloxacillin Sodium Imp. D (EP) IUPAC Name: 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid SMILES: Cc1onc(c1C(=O)O)c2c(F)cccc2Cl
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.