missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Fluoro-4-methylphenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 15408075
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408075 250 mg 250mg
15485087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408075 Supplier Thermo Scientific Alfa Aesar Supplier No. H32370.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 852181-17-0
Molecular Formula C10H11FO2
Molecular Weight (g/mol) 182.19
MDL Number MFCD09025394
InChI Key HQOAGAGIHCHXLP-UHFFFAOYSA-N
Synonym 3-2-fluoro-4-methylphenyl propionic acid, 3-2-fluoro-4-methylphenyl propanoic acid
PubChem CID 46737562
IUPAC Name 3-(2-fluoro-4-methylphenyl)propanoic acid
SMILES CC1=CC(F)=C(CCC(O)=O)C=C1

Spécification

Boiling Point 98°C to 100°C
Quantity 250 mg
Formula Weight 182.19
Percent Purity 97%
Chemical Name or Material 3-(2-Fluoro-4-methylphenyl)propionic acid
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.