missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10738275
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10738275 1 g 1g
10347124 250 mg 250mg
10051334 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10738275 Supplier Thermo Scientific Maybridge Supplier No. CC46104DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 85553-52-2
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD03990461
InChI Key BTAANNDAXIYWAN-UHFFFAOYSA-N
Synonym 3-2-furyl benzaldehyde, 3-furan-2-yl benzaldehyde, 3-furyl phenyl ketone, benzaldehyde, 3-2-furanyl, ketone, 3-furyl phenyl, 3-furan-2-yl-benzaldehyde, 3-furanylphenylmethanone, 3-furfanylphenylmethanone, 2-3-formylphenyl furan, 3-2-furanyl benzaidehyde
PubChem CID 201640
IUPAC Name 3-(furan-2-yl)benzaldehyde
SMILES O=CC1=CC=CC(=C1)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.