missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%

Product Code. 11452858
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452858 250 g 250g
11442858 50 g 50g
11432858 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452858 Supplier Thermo Scientific Alfa Aesar Supplier No. A16827.30
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-14-1 | C10H14O4 | 198.22 g/mol

3-(2-Methoxyphenoxy)-1,2-propanediol is involved in centrally acting muscle relaxant with expectorant properties. It is often used as anesthetic during animal surgery. It is used in the treatment of coughing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (50 mg/ml at 25°C), methanol, DMSO (40 mg/ml at 25°C), DMF, ethanol (50 mg/ml), glycerol and glycol.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-14-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00016873
InChI Key HSRJKNPTNIJEKV-UHFFFAOYNA-N
Synonym guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol
PubChem CID 3516
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
SMILES COC1=CC=CC=C1OCC(O)CO

Specifications

Melting Point 80°C to 82°C
Boiling Point 215°C (19 mmHg)
Quantity 250 g
Beilstein 2049375
Merck Index 14,4555
Solubility Information Soluble in water (50mg/ml at 25°C),methanol,DMSO (40mg/ml at 25°C),DMF,ethanol (50mg/ml),glycerol and glycol.
Formula Weight 198.22
Percent Purity ≥99%
Chemical Name or Material 3-(2-Methoxyphenoxy)-1,2-propanediol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.