missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Triphenylpropionic acid, 98%, Thermo Scientific™

Product Code. 11482678
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482678 25 g 25g
11472678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482678 Supplier Thermo Scientific Alfa Aesar Supplier No. A16686.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,3,3-Triphenylpropionic acid is used as pharmaceutical intermediates.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 900-91-4
Molecular Formula C21H18O2
Molecular Weight (g/mol) 302.373
MDL Number MFCD00002713
InChI Key XMSJLUKCGWQAHO-UHFFFAOYSA-N
Synonym 3,3,3-triphenylpropionic acid, tritylacetic acid, 3,3,3-triphenyl propanoic acid, 2-tritylacetic acid, pubchem12502, acmc-209r4v, 2-triphenylmethylacetic acid, 2-triphenylmethyl acetic acid, 3,3 3-triphenyl propionic acid
PubChem CID 136696
IUPAC Name 3,3,3-triphenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 180°C to 183°C
Quantity 25 g
Beilstein 2057448
Formula Weight 302.38
Percent Purity 98%
Chemical Name or Material 3,3,3-Triphenylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.