missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-(4-((Di-Boc)amino)-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)propanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(3-(4-((Di-Boc)amino)-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)propanoic Acid |
| Analyte or Component Names | 3-(3-(4-((Di-Boc)amino)-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)propanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H42 N6 O7 |
| InChI Formula | InChI=1S/C35H42N6O7/c1-34(2,3)47-32(44)40(33(45)48-35(4,5)6)30-28-29(23-14-16-26(17-15-23)46-25-12-8-7-9-13-25)38-41(31(28)37-22-36-30)24-11-10-19-39(21-24)20-18-27(42)43/h7-9,12-17,22,24H,10-11,18-21H2,1-6H3,(H,42,43) |
| Solution Type | Neat |
| SMILES | CC(C)(C)OC(=O)N(C(=O)OC(C)(C)C)c1ncnc2c1c(nn2C3CCCN(CCC(=O)O)C3)c4ccc(Oc5ccccc5)cc4 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 658.744 |
| Quantity | 500 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?