missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dichlorophenyl)-1,1-dimethylurea, 97%

Product Code. 11397915
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397915 50 g 50g
11307925 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11397915 Supplier Thermo Scientific Alfa Aesar Supplier No. L02986.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 330-54-1 | C9H10Cl2N2O | 233.092 g/mol

A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phenyl urea pre-emergent agrochemical. Also an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyestuff

Solubility
Soluble in water (0.049g/L), chloroform and methanol. Very slightly soluble in many organic solvents.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330-54-1
Molecular Formula C9H10Cl2N2O
Molecular Weight (g/mol) 233.092
MDL Number MFCD00018136
InChI Key XMTQQYYKAHVGBJ-UHFFFAOYSA-N
Synonym diuron, 3-3,4-dichlorophenyl-1,1-dimethylurea, dcmu, dynex, duran, dichlorfenidim, herbatox, vonduron, dailon, karmex
PubChem CID 3120
ChEBI CHEBI:116509
IUPAC Name 3-(3,4-dichlorophenyl)-1,1-dimethylurea
SMILES CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl

Specifications

Melting Point 157°C to 160°C
Density 1.48
Boiling Point 180°C to 190°C (decomposition)
Odor Odorless
Quantity 50 g
UN Number UN3077
Beilstein 2215168
Merck Index 14,3382
Solubility Information Soluble in water (0.049g/L),chloroform and methanol. Very slightly soluble in many organic solvents.
Formula Weight 233.1
Percent Purity 97%
Chemical Name or Material 3-(3,4-Dichlorophenyl)-1,1-dimethylurea
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-(3,4-Dichlorophenyl)-1,1-dimethylurea
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Diuron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.