missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, 98%

Product Code. 11489093
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489093 5 g 5g
11479093 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489093 Supplier Thermo Scientific Alfa Aesar Supplier No. A11164.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 175136-66-0 | C17H6F12O | 454.215 g/mol

3,3',5,5'-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetrakis(trifluoromethyl)benzophenone, is used as reactive dye intermediates.

Solubility
Sparingly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175136-66-0
Molecular Formula C17H6F12O
Molecular Weight (g/mol) 454.215
MDL Number MFCD00042474
InChI Key GATWMPGNBWPCIY-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetrakis trifluoromethyl benzophenone, bis 3,5-bis trifluoromethyl phenyl methanone, maybridge1_002937, acmc-1c7yy, bis 3,5-bis trifluoromethyl phenyl ketone, di 3,5-di trifluoromethyl phenyl methanone, bis 3,5-bis trifluoromethyl phenyl methanone #, 3,3/',5,5/'-tetrakis trifluoromethyl benzophenone
PubChem CID 519439
IUPAC Name bis[3,5-bis(trifluoromethyl)phenyl]methanone
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 139°C to 140°C
Quantity 5 g
Solubility Information Sparingly soluble in water.
Formula Weight 454.22
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetrakis (trifluoromethyl)benzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.