missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 98%

Product Code. 15480127
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480127 1 g 1g
15400137 2.5 g 2.5g
15490127 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480127 Supplier Thermo Scientific Alfa Aesar Supplier No. J60332.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 207738-08-7 | C16H22Cl2N2 | 313.27 g/mol

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

Solubility
Soluble in DMSO (∼1.7 mg/ml), and 1:300 DMSO:PBS (pH 7.2) (0.03 mg/ml).

Notes
Moisture and light sensitive. Store away from oxidizing agents, heat, light, acids, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207738-08-7
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27
MDL Number MFCD00150104
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride
PubChem CID 19836601
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride
SMILES Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Color White
Quantity 1 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in DMSO (∽1.7mg/ml),and 1:300 DMSO:PBS (pH 7.2) (0.03mg/ml).
Formula Weight 313.27
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.