missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 98%

Product Code. p-7041803
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
This item is not returnable. View return policy

Product Code. 15480127

Brand: Thermo Scientific Alfa Aesar J60332.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207738-08-7 | C16H22Cl2N2 | 313.27 g/mol

3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbenzidine dihydrochloride hydrate is a chromophore substrate in ELISA assays. It is a noncarcinogenic substitute for benzidine.

Solubility
Soluble in DMSO (∼1.7 mg/ml), and 1:300 DMSO:PBS (pH 7.2) (0.03 mg/ml).

Notes
Moisture and light sensitive. Store away from oxidizing agents, heat, light, acids, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207738-08-7
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27
MDL Number MFCD00150104
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride dihydrate, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride dihydrate, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, pubchem9044, tetramethylbenzidine dihydrate dihydrochloride
PubChem CID 19836601
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrate;dihydrochloride
SMILES Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Color White
Quantity 1 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in DMSO (∽1.7mg/ml),and 1:300 DMSO:PBS (pH 7.2) (0.03mg/ml).
Formula Weight 313.27
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.