missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine dihydrochloride, 99%

Product Code. p-7040467
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15405596

Brand: Thermo Scientific Alfa Aesar H60245.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64285-73-0 | C16H22Cl2N2 | 313.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64285-73-0
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.27
MDL Number MFCD00012961
InChI Key NYNRGZULARUZCC-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine dihydrochloride, tmb dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethyl-, dihydrochloride, tetramethylbenzidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3',5,5'-tetramethyl-, hydrochloride 1:2, 4-4-amino-3,5-dimethylphenyl-2,6-dimethylaniline dihydrochloride, 3,3',5,5'-tetramethylbenzidine dihydrochloride hydrate, 3,3',5,5'-tetramethyl 1,1'-biphenyl-4,4'-diamine dihydrochloride
PubChem CID 174033
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1

Specifications

Melting Point 300°C
Quantity 250 mg
Sensitivity Light and moisture sensitive
Formula Weight 313.27
Percent Purity 99%
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.