missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, high sensitivity, Thermo Scientific Chemicals

Product Code. 15430187
Click to view available options
Quantity:
125 mL
500 mL
1000 mL
Unit Size:
1000mL
125mL
500mL
This item is not returnable. View return policy

Product Code. 15430187

Brand: Thermo Scientific Alfa Aesar J61325.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,3’,5,5’-Tetramethylbenzidine (TMB, CAS # 54827-17-7) is a blue chromogenic substance used in staining techniques such as immunohistochemistry.

TMB is a substrate for horseradish peroxidase. Recommended for ELISA (microwell) procedures,3,3?,5,5?-Tetramethylbenzidine has been used in a study to demonstrate that by measuring substrate absorption values off the absorption maximum, the measuring range of any immunoenzymometric assay can be extended significantly without jeopardizing assay characteristics. 3,3?,5,5?-Tetramethylbenzidine has also been used in a study to understand the formation of CuS concave superstructures with peroxidase-like activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• TMB is an aromatic amine that acts as a hydrogen donor in horseradish peroxidase-catalyzed reactions
• The transfer of hydrogen results in a blue stain associated with the target of interest
• This pH-dependent color change can be confirmed using spectrophotometric methods
• TMB is commonly used in immunohistochemistry; TMB is light-sensitive, and all applications should be performed in the dark

Application

• TMB is suitable for immunohistochemistry and immunocytochemistry applications including ELISA and cell and tissue staining

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Color Yellow
Quantity 500 mL
UN Number UN2810
Beilstein 2808541
Solubility Information Insoluble in water. Soluble in organic solvents such as DMSO,96 % Ethanol,Acetone,Chloroform,Toluene.
Formula Weight 240.35
Physical Form Liquid
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, high sensitivity

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.