missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Chloromethyl)phenoxy]-N,N-dimethylpropylamine hydrochloride, 95%, Thermo Scientific™

Product Code. 10403034
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10403034

Brand: Thermo Scientific Maybridge CC59474CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 926921-62-2
Molecular Formula C12H19Cl2NO
Molecular Weight (g/mol) 264.19
MDL Number MFCD09702394
InChI Key MCRYCZSEVCPYAW-UHFFFAOYSA-N
Synonym 3-3-chloromethyl phenoxy-n,n-dimethylpropylamine hydrochloride, 3-3-dimethylamino propoxy benzyl chloride hydrochloride, 3-3-chloromethyl phenoxy propyl dimethylamine hydrochloride, 3-3-chloromethyl phenoxy propyl dimethylamine, chloride, 3-3-chloromethyl phenoxy-n,n-dimethylpropan-1-amine hydrochloride, 1-propanamine,3-3-chloromethyl phenoxy-n,n-dimethyl-,hydrochloride 1:1, 3-3-chloromethyl phenoxy-n,n-dimethylpropan-1-amine-hydrogen chloride 1/1
PubChem CID 24229607
SMILES Cl.CN(C)CCCOC1=CC=CC(CCl)=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 3-[3-(chloromethyl)phenoxy]-N,N-dimethylpropan-1-amine;hydrochloride
Formula Weight 264.19
Percent Purity 95%
Chemical Name or Material 3-[3-(Chloromethyl)phenoxy]-N, N-dimethylpropylamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.