missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Chlorophenyl)-1,1-dimethylurea, TRC

Product Code. 30135840
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135840 1 g 1g
30133962 500 mg 500mg
30010025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135840 Supplier Toronto Research Chemicals Supplier No. TRCC6097851G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-34-8
Molecular Formula C9H11ClN2O
Molecular Weight (g/mol) 198.65
Synonym 3-(m-chlorophenyl)-1,1-dimethyl-Urea, N'-(3-Chlorophenyl)-N,N-dimethylurea, 1-(3-Chlorophenyl)-3,3-dimethylurea, 1-m-Chlorophenyl-3,3-dimethylurea, C 2034 N,N-Dimethyl-N'-3-chlorophenylurea, N-(3-Chlorophenyl)-N',N'-dimethylurea, N-(m-Chlorophenyl)-N',N'-dimethylurea
IUPAC Name 3-(3-chlorophenyl)-1,1-dimethylurea
SMILES O=C(N(C)C)NC1=CC(Cl)=CC=C1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H11ClN2O/c1-12(2)9(13)11-8-5-3-4-7(10)6-8/h3-6H,1-2H3,(H,11,13)
Formula Weight 198.056
Chemical Name or Material 3-(3-Chlorophenyl)-1,1-dimethylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.