missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzophenone, 90%

Product Code. 11327246
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327246 5 g 5g
11317246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327246 Supplier Thermo Scientific Alfa Aesar Supplier No. L09128.06
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 611-79-0 | C13H12N2O | 212.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-79-0
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00014774
InChI Key TUQQUUXMCKXGDI-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzophenone, bis 3-aminophenyl methanone, methanone, bis 3-aminophenyl, di3-aminophenyl ketone, 3,3'-diaminodiphenyl ketone, pubchem3381, acmc-1awhc, cbmicro_014188, 3,3'-diamino benzophenone, 3-3-aminobenzoyl aniline
PubChem CID 69145
IUPAC Name bis(3-aminophenyl)methanone
SMILES NC1=CC(=CC=C1)C(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 148°C to 152°C
Boiling Point 285°C (11 mmHg)
Quantity 5 g
Beilstein 2837605
Formula Weight 212.25
Percent Purity ≥90%
Chemical Name or Material 3,3'-Diaminobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.