missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminodiphenyl sulfone, 98%

Product Code. 11327756
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327756 25 g 25g
11337756 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327756 Supplier Thermo Scientific Alfa Aesar Supplier No. L11400.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 599-61-1 | C12H12N2O2S | 248.30 g/mol

3,3'-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminodiphenyl sulfone is used for curing agent for epoxy resin and also important monomer for polysulfonamide fiber(PSA).

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-61-1
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.30
MDL Number MFCD00007792
InChI Key LJGHYPLBDBRCRZ-UHFFFAOYSA-N
Synonym 3,3'-sulfonyldianiline, 3,3'-diaminodiphenyl sulfone, 3-aminophenyl sulfone, benzenamine, 3,3'-sulfonylbis, 3,3'-sulphonyldianiline, bis m-aminophenyl sulfone, 3-aminophenyl-sulfone, 3,3'-diaminophenyl sulfone, 3,3'-sulfonylbis aniline, sulfone, bis m-aminophenyl
PubChem CID 11741
IUPAC Name 3-(3-aminophenyl)sulfonylaniline
SMILES NC1=CC(=CC=C1)S(=O)(=O)C1=CC(N)=CC=C1

Specifications

Melting Point 171°C to 174°C
Odor Odorless
Quantity 25 g
Beilstein 2215938
Solubility Information Insoluble in water.
Formula Weight 248.3
Percent Purity 98%
Chemical Name or Material 3,3'-Diaminodiphenyl sulfone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.