missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diindolylmethane, MedChemExpress

Product Code. 16486916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486916 100 mg 100mg
16461196 200 mg 200mg
16431786 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486916 Supplier Medchem Express Supplier No. HY15758
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 3,3'-Diindolylmethane is a strong, pure androgen receptor (AR) antagonist.

3,3'-Diindolylmethane is a strong, pure androgen receptor (AR) antagonist.

Specifications

Chemical Name or Material 3,3'-Diindolylmethane
Physical Form Solid
CAS 1968-05-4
Color White to Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H14N2
For Use With (Application) Cancer-programmed cell death
Synonym DIM Arundine HB 236
Solubility Information DMSO : ≥ 100 mg/mL (405.99 mM)
SMILES C1(CC2=CNC3=C2C=CC=C3)=CNC4=C1C=CC=C4
Molecular Weight (g/mol) 246.31
Formula Weight 246.31
Percent Purity 98.09%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.