missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethoxybenzidine dihydrochloride, 98%

Product Code. 10167303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10167303 5 g Glass bottle 5g
10553551 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167303 Supplier Thermo Scientific Acros Supplier No. 184470050
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20325-40-0 | C14H16N2O2·2HCl | 317.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20325-40-0
Molecular Formula C14H16N2O2·2HCl
Molecular Weight (g/mol) 317.21
MDL Number MFCD00012488
InChI Key UXTIAFYTYOEQHV-UHFFFAOYSA-N
Synonym 3,3'-dimethoxybenzidine dihydrochloride, o-dianisidine dihydrochloride, 3,3'-dimethoxy-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-qv96qa6ukk, dianisidine dihydrochloride, ccris 998, o-dianisidine hcl, qv96qa6ukk, c.i. disperse black 6 dihydrochloride
PubChem CID 62311
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline;dihydrochloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)OC)N.Cl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 268.0°C
Color Beige to Gray-Purple
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 13, 807
Formula Weight 317.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3'-Dimethoxybenzidine dihydrochloride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,3''-Dimethoxybenzidine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3,3''-Dimethoxybenzidine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.