missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropionitrile, Thermo Scientific Chemicals

Product Code. 16637062 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 16637062

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2286-54-6
MDL Number MFCD00129747

Specifications

Alpha Vector DIPHENYLPROPIONITRILE 010
Melting Point 86°C to 89°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C15 H13 N
Quantity 5 g
Solubility Information soluble in hot methanol
Molecular Weight (g/mol) 207.27
Formula Weight 207.27
Physical Form Crystalline Powder
Chemical Name or Material 3,3-Diphenylpropionitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.