missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Diphenylpropionitrile, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Alpha Vector | DIPHENYLPROPIONITRILE 010 |
| Melting Point | 86°C to 89°C |
| Color | White to Yellow |
| Infrared Spectrum | Conforms |
| Packaging | glass bottle |
| Molecular Formula | C15 H13 N |
| Quantity | 5 g |
| Solubility Information | soluble in hot methanol |
| Molecular Weight (g/mol) | 207.27 |
| Formula Weight | 207.27 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction