missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dithiobis[6-nitrobenzoic Acid], TRC

Product Code. 30011155
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011155 5 g 5g
30007741 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011155 Supplier Toronto Research Chemicals Supplier No. TRCD4798405G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-78-3
Molecular Formula C14H8N2O8S2
Molecular Weight (g/mol) 396.35
Synonym 2,2'-Dinitro-5,5'-dithiodibenzoic Acid, 3,3'-Dithiobis(6-nitrobenzoic Acid), 5,5'-Dithiobis[2-nitrobenzoic Acid], Ba 2767, DTNB, Ellman’s Reagent
IUPAC Name 5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
SMILES OC(=O)c1cc(SSc2ccc(c(c2)C(=O)O)[N+](=O)[O-])ccc1[N+](=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H8N2O8S2/c17-13(18)9-5-7(1-3-11(9)15(21)22)25-26-8-2-4-12(16(23)24)10(6-8)14(19)20/h1-6H,(H,17,18)(H,19,20)
Formula Weight 395.9722
Chemical Name or Material 3,3'-Dithiobis[6-nitrobenzoic Acid]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.