missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methoxyphenyl)-1H-pyrazole, 97%

Product Code. 11335469
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335469 1 g 1g
11345469 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335469 Supplier Thermo Scientific Alfa Aesar Supplier No. H50435.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 144026-74-4 | C10H10N2O | 174.203 g/mol

3-(3-Methoxyphenyl)-1H-pyrazole is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Methoxyphenyl)-1H-pyrazole is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144026-74-4
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD01940432
InChI Key JAZQIOZMPNVSII-UHFFFAOYSA-N
Synonym 3-3-methoxyphenyl-1h-pyrazole, 5-3-methoxyphenyl-1h-pyrazole, 1h-pyrazole, 3-3-methoxyphenyl, 3-3-methoxyphenyl pyrazole, 3-3-methoxy-phenyl-1h-pyrazole, 3-3-methoxyphenyl-2h-pyrazole, acmc-20ankv, 1-methoxy-3-pyrazol-3-ylbenzene, 3-methoxy-1-pyrazol-3-ylbenzene, 1h-pyrazole,3-3-methoxyphenyl
PubChem CID 2736766
IUPAC Name 5-(3-methoxyphenyl)-1H-pyrazole
SMILES COC1=CC=CC(=C1)C2=CC=NN2

Specifications

Melting Point 90°C to 91°C
Quantity 1 g
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material 3-(3-Methoxyphenyl)-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.