missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(2-Methoxyethyl)phenoxy]-1,2-propanediol, TRC

Product Code. 30137291
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137291 250 mg 250mg
30004706 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137291 Supplier Toronto Research Chemicals Supplier No. TRCM226140250MG
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62572-90-1
Molecular Formula C12 H18 O4
Molecular Weight (g/mol) 226.27
Synonym (2RS)-3-[4-(2-Methoxyethyl)phenoxy]propane-1,2-diol, Metoprolol Succinate Imp. D (EP), Metoprolol Tartrate Imp. D (EP)
IUPAC Name 3-[4-(2-methoxyethyl)phenoxy]propane-1,2-diol
SMILES COCCc1ccc(OCC(O)CO)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H18O4/c1-15-7-6-10-2-4-12(5-3-10)16-9-11(14)8-13/h2-5,11,13-14H,6-9H2,1H3
Formula Weight 226.1205
Chemical Name or Material 3-[4-(2-Methoxyethyl)phenoxy]propane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.