missing translation for 'onlineSavingsMsg'
Learn More

3',4',5,7-Tetramethoxyflavone, 97%

Product Code. 11323556
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323556 100 mg 100mg
11313556 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323556 Supplier Thermo Scientific Alfa Aesar Supplier No. L14496.MC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 855-97-0 | C19H18O6 | 342.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855-97-0
Molecular Formula C19H18O6
Molecular Weight (g/mol) 342.35
MDL Number MFCD00017558
InChI Key CLXVBVLQKLQNRQ-UHFFFAOYSA-N
Synonym 3',4',5,7-tetramethoxyflavone, luteolin tetramethyl ether, tetramethoxyluteolin, 5,7,3',4'-tetramethylluteolin, tetramethylluteolin, 2-3,4-dimethoxyphenyl-5,7-dimethoxy-4h-chromen-4-one, luteolin tetramethylether, 2-3,4-dimethoxyphenyl-5,7-dimethoxychromen-4-one, luteolin 5,7,3',4'-tetramethyl ether, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-5,7-dimethoxy
PubChem CID 631170
IUPAC Name 2-(3,4-dimethoxyphenyl)-5,7-dimethoxychromen-4-one
SMILES COC1=CC(OC)=C2C(=O)C=C(OC2=C1)C1=CC=C(OC)C(OC)=C1

Specifications

Melting Point 193°C to 197°C
Quantity 100 mg
Beilstein 336320
Formula Weight 342.35
Percent Purity 97%
Chemical Name or Material 3',4',5,7-Tetramethoxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.