missing translation for 'onlineSavingsMsg'
Learn More

3-(4,5-Diphenyl-1,3-oxazol-2-yl)propanoic acid, 97%, Thermo Scientific™

Product Code. 10742133
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10742133 1 g 1g
10703377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10742133 Supplier Thermo Scientific Maybridge Supplier No. SS00042DA
 Restricted Item
Estimated Shipment: 18-01-2027
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Kemiske identifikatorer

CAS 21256-18-8
Molecular Formula C18H15NO3
Molecular Weight (g/mol) 293.32
MDL Number MFCD00215977
InChI Key OFPXSFXSNFPTHF-UHFFFAOYSA-N
Synonym oxaprozin, daypro, alvo, 3-4,5-diphenyl-1,3-oxazol-2-yl propanoic acid, oxaprozine, 3-4,5-diphenyloxazol-2-yl propanoic acid, deflam, oxaprozina, oxaprozinum, voir
PubChem CID 4614
ChEBI CHEBI:7822
IUPAC Name 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
SMILES OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Tekniske data

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Merck Index 15, 7023
Formula Weight 293.32
Percent Purity 97%
Chemical Name or Material 3-(4, 5-Diphenyl-1, 3-oxazol-2-yl)propanoic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.