missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzaldehyde, 98%, Thermo Scientific™

Product Code. 10422903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
Product Code.
10422903 1g; Glass bottle
1 options
This item is not returnable. View return policy

Product Code. 10422903

Brand: Thermo Scientific Acros 384210010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 132123-54-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 25.3°C
Color Colorless to Yellow
Density 1.4200g/mL
Boiling Point 174.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H3F3O
Linear Formula F3C6H2CHO
MDL Number MFCD00083523
Quantity 1 g
Specific Gravity 1.42
Synonym benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci
Solubility Information Solubility in water: 2.2g/L (20°C)
InChI Key NLAVHUUABUFSIG-UHFFFAOYSA-N
SMILES FC1=CC(C=O)=CC(F)=C1F
IUPAC Name 3,4,5-trifluorobenzaldehyde
Molecular Weight (g/mol) 160.10
PubChem CID 2776954
Formula Weight 160.09
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 3, 4, 5-Trifluorobenzaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4,5-Trifluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.