missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11334066
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334066 250 mg 250mg
11314066 1 g 1g
11324066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334066 Supplier Thermo Scientific Alfa Aesar Supplier No. L19188.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177787-26-7
Molecular Formula C7H2ClF3O
Molecular Weight (g/mol) 194.537
MDL Number MFCD00083526
InChI Key YRUNCQNKZIQTEO-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzoylchloride, benzoyl chloride,3,4,5-trifluoro, acmc-20ange, pubchem20778, 3,4,5-trifluorobenzoic acid chloride, 3,4,5-tris fluoranyl benzoyl chloride, 3,4,5-trifluorobenzoyl chloride, benzoyl chloride, 3,4,5-trifluoro-9ci
PubChem CID 2777023
IUPAC Name 3,4,5-trifluorobenzoyl chloride
SMILES C1=C(C=C(C(=C1F)F)F)C(=O)Cl

Specifications

Refractive Index 1.4898
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 194.54
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzoyl chloride
compliance-icons
Product Identifier
  • 3,4,5-Trifluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.