missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorophenol, 97%, Thermo Scientific™

Product Code. 11400148
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11400148

Brand: Thermo Scientific Alfa Aesar B23718.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99627-05-1
Molecular Formula C6H3F3O
Molecular Weight (g/mol) 148.084
MDL Number MFCD00042427
InChI Key ZRTWIJKGTUGZJY-UHFFFAOYSA-N
Synonym phenol, 3,4,5-trifluoro, 3,4,5-trifluoro-phenol, pubchem2316, 3,4,5-trifluoropheno, acmc-209sdi, ksc486m6r, 3 , 4 , 5-trifluorophenol, phenol, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenol
PubChem CID 2777943
IUPAC Name 3,4,5-trifluorophenol
SMILES C1=C(C=C(C(=C1F)F)F)O

Specifications

Melting Point 52°C to 56°C
Boiling Point 177°C
Quantity 1 g
UN Number UN3261
Beilstein 8830884
Formula Weight 148.09
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.