missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorophenylboronic acid, 95+%, Thermo Scientific™

Product Code. 10078951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10078951 250 mg Glass bottle 250mg
10039041 1 g Glass bottle 1g
10509813 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10078951 Supplier Thermo Scientific Acros Supplier No. 368192500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143418-49-9
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.901
MDL Number MFCD02093069
InChI Key UHDDEIOYXFXNNJ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj
PubChem CID 2734671
IUPAC Name (3,4,5-trifluorophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)F)F)F)(O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 95+%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 175.9
Percent Purity ≥95.0%
Physical Form Powder
Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,4,5-Trifluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.