missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzaldehyde hydrate, 97%

Product Code. 11449536
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449536 5 g 5g
11439536 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449536 Supplier Thermo Scientific Alfa Aesar Supplier No. A19270.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13677-79-7 | C7H6O4 | 154.12 g/mol

3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.

Solubility
Soluble in DMSO and Methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13677-79-7
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00003371
InChI Key RGZHEOWNTDJLAQ-UHFFFAOYSA-N
Synonym gallaldehyde, benzaldehyde, 3,4,5-trihydroxy, pyrogallol-5-carboxaldehyde, 3,4,5-trihydroxy benzaldehyde, pubchem21300, 3,4,5-trihydroxybenz, acmc-209c9s, 3,4,5-trihydroxybenzaldhyde, 3,4,5-trihydroxy-benzaldehyde, 3,4,5-tris oxidanyl benzaldehyde
PubChem CID 83651
IUPAC Name 3,4,5-trihydroxybenzaldehyde
SMILES OC1=CC(C=O)=CC(O)=C1O

Specifications

Melting Point ∼212°C (decomposition)
Quantity 5 g
Beilstein 2091654
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO and Methanol.
Formula Weight 154.12 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 3,4,5-Trihydroxybenzaldehyde hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.