missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyaniline, 97%

Product Code. 10316690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10316690 10 g Glass bottle 10g
10225240 25 g Glass bottle 25g
10135833 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316690 Supplier Thermo Scientific Acros Supplier No. 139900100
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24313-88-0 | C9H13NO3 | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24313-88-0
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.21
MDL Number MFCD00008393
InChI Key XEFRNCLPPFDWAC-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
PubChem CID 32285
IUPAC Name 3,4,5-trimethoxyaniline
SMILES COC1=CC(=CC(=C1OC)OC)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 110.0°C to 113.0°C
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3O)3C6H2NH2
Quantity 10 g
Beilstein 13, 826
Solubility Information Solubility in water: insoluble
Formula Weight 183.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3, 4, 5-Trimethoxyaniline
Show More Show Less
compliance-icons
Product Identifier
  • 3,4,5-Trimethoxyaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.