missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyaniline, 98+%

Product Code. p-7043617
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
missing translation for 'unitSize'
25g
5g
This item is not returnable. View return policy

Product Code. 11371347

missing translation for 'mfr': Thermo Scientific Alfa Aesar L01619.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24313-88-0 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24313-88-0
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00008393
InChI Key XEFRNCLPPFDWAC-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
PubChem CID 32285
IUPAC Name 3,4,5-trimethoxyaniline
SMILES COC1=CC(=CC(=C1OC)OC)N

Specifications

Melting Point 110°C to 113°C
Quantity 5 g
UN Number UN2811
Beilstein 642919
Formula Weight 183.21
Percent Purity ≥98%
Chemical Name or Material 3,4,5-Trimethoxyaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.