missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzaldehyde, 99%

Product Code. 10742875 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10742875

Brand: Thermo Scientific Acros 139915000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-81-7 | C10H12O4 | 196.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-81-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.2
MDL Number MFCD00003364
InChI Key OPHQOIGEOHXOGX-UHFFFAOYSA-N
PubChem CID 6858
IUPAC Name 3,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1OC)OC)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 73.0°C to 76.0°C
Color Cream to White
Boiling Point 163.0°C to 165.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CHO
Quantity 500 g
Beilstein 08, 391
Synonym benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,toluene and acetone
Formula Weight 196.2
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material 3, 4, 5-Trimethoxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 3,4,5-Trimethoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.