missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzyl alcohol, 98%

Product Code. 11464384
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464384 50 g 50g
11454384 10 g 10g
11474384 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464384 Supplier Thermo Scientific Alfa Aesar Supplier No. A13071.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3840-31-1 | C10H14O4 | 198.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3840-31-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00004639
InChI Key QPHLRCUCFDXGLY-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzyl alcohol, 3,4,5-trimethoxyphenyl methanol, benzenemethanol, 3,4,5-trimethoxy, 3,4,5-trimethoxybenzylalcohol, 3,4,5-trimethoxybenzylic alcohol, benzyl alcohol, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methan-1-ol, acmc-1cp7y, 3,4,5-trimethoxybenzalcohol, ksc494s0n
PubChem CID 77449
IUPAC Name (3,4,5-trimethoxyphenyl)methanol
SMILES COC1=CC(=CC(=C1OC)OC)CO

Specifications

Density 1.199
Boiling Point 228°C (25 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5445
Quantity 50 g
Beilstein 2104523
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.