missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzylamine, 96%

Product Code. 11393077
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393077 5 g 5g
11303087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393077 Supplier Thermo Scientific Alfa Aesar Supplier No. L13461.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18638-99-8 | C10H15NO3 | 197.234 g/mol

3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18638-99-8
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00017150
InChI Key YUPUSBMJCFBHAP-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
PubChem CID 87736
IUPAC Name (3,4,5-trimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1OC)OC)CN

Specifications

Density 1.155
Boiling Point 120°C to 122°C (0.05 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.548
Quantity 5 g
UN Number UN2735
Beilstein 1107570
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 197.24
Percent Purity 96%
Chemical Name or Material 3,4,5-Trimethoxybenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.